Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Pyridineacetonitrile hydrochloride, 98%

Click to view available options
Quantity:
1g
5g
25g
92333-25-0
C7H7ClN2
154.597
MFCD00012825
UBYQSEXKPQZPCN-UHFFFAOYSA-N
4-pyridylacetonitrile hydrochloride,2-pyridin-4-yl acetonitrile hydrochloride,4-pyridyl acetonitrile hydrochloride,4-pyridylacetonitrile hcl,4-pyridineacetonitrile hydrochloride,pyridine-4-acetonitrile hydrochloride,4-pyridineacetonitrile, monohydrochloride,4-pyridineacetonitrilehcl,4-cyanomethylpyridine hydrochloride,4-pyridylaceto-nitrile hydrochloride
12396608
2-pyridin-4-ylacetonitrile;hydrochloride
C1=CN=CC=C1CC#N.Cl
92333-25-0
C7H7ClN2
154.597
MFCD00012825
UBYQSEXKPQZPCN-UHFFFAOYSA-N
4-pyridylacetonitrile hydrochloride,2-pyridin-4-yl acetonitrile hydrochloride,4-pyridyl acetonitrile hydrochloride,4-pyridylacetonitrile hcl,4-pyridineacetonitrile hydrochloride,pyridine-4-acetonitrile hydrochloride,4-pyridineacetonitrile, monohydrochloride,4-pyridineacetonitrilehcl,4-cyanomethylpyridine hydrochloride,4-pyridylaceto-nitrile hydrochloride
12396608
2-pyridin-4-ylacetonitrile;hydrochloride
C1=CN=CC=C1CC#N.Cl
Melting Point ~260°C (decomposition)
Quantity 25g
UN Number UN3439
Formula Weight 154.6
Percent Purity 98%
Chemical Name or Material 4-Pyridineacetonitrile hydrochloride

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.