Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyridinecarboxaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
872-85-5
C6H5NO
107.11
MFCD00006425
BGUWFUQJCDRPTL-UHFFFAOYSA-N
4-pyridinecarboxaldehyde,isonicotinaldehyde,4-formylpyridine,isonicotinic aldehyde,p-pyridinealdehyde,pyridine-4-carboxaldehyde,4-pyridinealdehyde,4-pyridylaldehyde,4-pyridinecarbaldehyde,pyridine-4-aldehyde
13389
pyridine-4-carbaldehyde
C1=CN=CC=C1C=O
872-85-5
C6H5NO
107.11
MFCD00006425
BGUWFUQJCDRPTL-UHFFFAOYSA-N
4-pyridinecarboxaldehyde,isonicotinaldehyde,4-formylpyridine,isonicotinic aldehyde,p-pyridinealdehyde,pyridine-4-carboxaldehyde,4-pyridinealdehyde,4-pyridylaldehyde,4-pyridinecarbaldehyde,pyridine-4-aldehyde
13389
pyridine-4-carbaldehyde
C1=CN=CC=C1C=O
Flash Point 82°C
Specific Gravity 1.138
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5420 to 1.5450
Quantity 100g
Color Yellow to Brown
Density 1.1380g/mL
Boiling Point 77.0°C to 78.0°C (12.0mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula (C5H4N)COH
Fieser 11,448
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: miscible with ethanol,acetone,toluene
Formula Weight 107.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Pyridinecarboxaldehyde
Show More Show Less

GHS H Statement
Harmful if inhaled.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 212-832-3