Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyridinecarboxaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
872-85-5
C6H5NO
107.11
MFCD00006425
BGUWFUQJCDRPTL-UHFFFAOYSA-N
4-pyridinecarboxaldehyde,isonicotinaldehyde,4-formylpyridine,isonicotinic aldehyde,p-pyridinealdehyde,pyridine-4-carboxaldehyde,4-pyridinealdehyde,4-pyridylaldehyde,4-pyridinecarbaldehyde,pyridine-4-aldehyde
13389
pyridine-4-carbaldehyde
C1=CN=CC=C1C=O
872-85-5
C6H5NO
107.11
MFCD00006425
BGUWFUQJCDRPTL-UHFFFAOYSA-N
4-pyridinecarboxaldehyde,isonicotinaldehyde,4-formylpyridine,isonicotinic aldehyde,p-pyridinealdehyde,pyridine-4-carboxaldehyde,4-pyridinealdehyde,4-pyridylaldehyde,4-pyridinecarbaldehyde,pyridine-4-aldehyde
13389
pyridine-4-carbaldehyde
C1=CN=CC=C1C=O
Flash Point 82°C
Specific Gravity 1.138
Packaging Glass bottle
Quantity 500g
Infrared Spectrum Authentic
Density 1.1380g/mL
Refractive Index 1.5420 to 1.5450
Color Yellow to Brown
Boiling Point 77.0°C to 78.0°C (12.0mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C5H4N)COH
Fieser 11,448
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: miscible with ethanol,acetone,toluene
Formula Weight 107.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Pyridinecarboxaldehyde
Show More Show Less

GHS H Statement:
Harmful if inhaled.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger