Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (4-Pyridylthio)acetic acid, 98%

Click to view available options
Quantity:
25g
100g
500g
10351-19-6
C7H7NO2S
169.198
MFCD00006424
PGUPJAPHYIEKLT-UHFFFAOYSA-N
4-pyridylthio acetic acid,2-pyridin-4-ylthio acetic acid,acetic acid, 4-pyridinylthio,4-pyridylthioacetic acid,pyridin-4-ylsulfanyl acetic acid,2-pyridin-4-ylsulfanyl acetic acid,4-pyridinylthio acetic acid,acetic acid, 2-4-pyridinylthio,s-4-pyridyl thioglycolic acid,2-4-pyridylthio acetic acid
82574
2-pyridin-4-ylsulfanylacetic acid
C1=CN=CC=C1SCC(=O)O
10351-19-6
C7H7NO2S
169.198
MFCD00006424
PGUPJAPHYIEKLT-UHFFFAOYSA-N
4-pyridylthio acetic acid,2-pyridin-4-ylthio acetic acid,acetic acid, 4-pyridinylthio,4-pyridylthioacetic acid,pyridin-4-ylsulfanyl acetic acid,2-pyridin-4-ylsulfanyl acetic acid,4-pyridinylthio acetic acid,acetic acid, 2-4-pyridinylthio,s-4-pyridyl thioglycolic acid,2-4-pyridylthio acetic acid
82574
2-pyridin-4-ylsulfanylacetic acid
C1=CN=CC=C1SCC(=O)O
Melting Point ~260°C (decomposition)
Quantity 25g
Beilstein 129738
Formula Weight 169.2
Percent Purity 98%
Chemical Name or Material (4-Pyridylthio)acetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA