Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ (4-Pyridylthio)acetic acid, 98%
Click to view available options
Quantity:
25g
100g
500g
Specifications
Quantity | 500g |
Melting Point | ~260°C (decomposition) |
Beilstein | 129738 |
Formula Weight | 169.2 |
Percent Purity | 98% |
Chemical Name or Material | (4-Pyridylthio)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 233-765-6
TSCA : Yes
Recommended Storage : Ambient temperatures