Alfa Aesar™ 4-Sulfamoylbenzoic acid, 96%

Manufacturer: Alfa Aesar™ B2244930

Quantity 250g

 View more versions of this product

Catalog No. AAB2244930

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 138-41-0
    Molecular Formula C7H7NO4S
    Formula Weight 201.2
    MDL Number MFCD00007938
    Beilstein 1875393
    Merck Index 14,1876
    Synonym 4-(Aminosulfonyl)benzoic acid; 4-Carboxybenzenesulfonamide
    Chemical Name or Material 4-Sulfamoylbenzoic acid

    Specifications

    Formula Weight 201.2
    TSCA TSCA
    Beilstein 1875393
    Synonym 4-(Aminosulfonyl)benzoic acid; 4-Carboxybenzenesulfonamide
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈285°C (dec.)
    Quantity 250g
    Molecular Formula C7H7NO4S
    MDL Number MFCD00007938
    CAS 138-41-0
    Merck Index 14,1876
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number DH6820000
    Chemical Name or Material 4-Sulfamoylbenzoic acid
    Assay 96%

    Description

    Applications
    Sulfamoylbenzoic acid is a moiety of sulfamyl diuretics or antihypertensive. It is used as an antiviral and an intermediate for the manufacture of frusemide which used as a diuretic and as an antihypertensive.

    Solubility
    Insoluble in water.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit