Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Amylcalix[8]arene, 95%, Acros Organics

Click to view available options
Quantity:
1g
93503-77-6
C96H128O8
1410.072
MFCD01863233
NSLJZLPZJOKHDZ-UHFFFAOYSA-N
4-tert-amylcalix 8 arene,4-tert-amylcalix 8 arene
2733650
CCC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=CC(=CC(=C6O)CC7=C(C(=CC(=C7)C(C)(C)CC)CC8=C(C(=CC(=C8)C(C)(C)CC)CC9=C(C(=CC(=C9)C(C)(C)CC)C2)O)O)O)C(C)(C)CC)C(C)(C)CC)C(C)(C)CC)C(C)(C)CC)O
93503-77-6
C96H128O8
1410.072
MFCD01863233
NSLJZLPZJOKHDZ-UHFFFAOYSA-N
4-tert-amylcalix 8 arene,4-tert-amylcalix 8 arene
2733650
CCC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=CC(=CC(=C6O)CC7=C(C(=CC(=C7)C(C)(C)CC)CC8=C(C(=CC(=C8)C(C)(C)CC)CC9=C(C(=CC(=C9)C(C)(C)CC)C2)O)O)O)C(C)(C)CC)C(C)(C)CC)C(C)(C)CC)C(C)(C)CC)O
Chelation/Complexation reagent
Infrared Spectrum Authentic
Quantity 1g
Color White to Beige
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 1410.1
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material 4-tert-Amylcalix[8]arene

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.