Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(tert-Butoxy)phenol, 98%

Click to view available options
Quantity:
1g
5g
2460-87-9
C10H14O2
166.22
MFCD02183556
CIICLJLSRUHUBY-UHFFFAOYSA-N
4-tert-butoxy phenol,4-tert-butoxyphenol,p-tert-butoxyphenol,4-t-butoxyphenol,phenol, 4-1,1-dimethylethoxy,unii-4w56a32ou8,4-2-methylpropan-2-yl oxy phenol,phenol, p-tert-butoxy,4-tert-butoxy-phenol,acmc-209gdf
2773621
4-[(2-methylpropan-2-yl)oxy]phenol
CC(C)(C)OC1=CC=C(C=C1)O
2460-87-9
C10H14O2
166.22
MFCD02183556
CIICLJLSRUHUBY-UHFFFAOYSA-N
4-tert-butoxy phenol,4-tert-butoxyphenol,p-tert-butoxyphenol,4-t-butoxyphenol,phenol, 4-1,1-dimethylethoxy,unii-4w56a32ou8,4-2-methylpropan-2-yl oxy phenol,phenol, p-tert-butoxy,4-tert-butoxy-phenol,acmc-209gdf
2773621
4-[(2-methylpropan-2-yl)oxy]phenol
CC(C)(C)OC1=CC=C(C=C1)O
Melting Point 151°C to 153°C
Quantity 1g
Odor Phenol-like
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 4-(tert-Butoxy)phenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures