Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
330794-35-9
C18H28BNO4
333.24
CUDCEJRRWNIPDL-UHFFFAOYSA-N
4-n-boc-aminomethyl phenylboronic acid pinacol ester,tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate,4-n-boc-amino methyl phenylboronic acid pinacol ester,4-tert-butoxycarbonylaminomethyl phenylboronic acid, pinacol ester,tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl carbamate,4-tert-butoxycarbonylaminomethyl phenylboronic acid pinacol ester,4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-carbamic acid tert-butyl ester,pubchem5247,4-n-boc-aminomethyl phenylboronic acid,pinacol ester
3415442
tert-butyl N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CNC(=O)OC(C)(C)C
330794-35-9
C18H28BNO4
333.24
CUDCEJRRWNIPDL-UHFFFAOYSA-N
4-n-boc-aminomethyl phenylboronic acid pinacol ester,tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate,4-n-boc-amino methyl phenylboronic acid pinacol ester,4-tert-butoxycarbonylaminomethyl phenylboronic acid, pinacol ester,tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl carbamate,4-tert-butoxycarbonylaminomethyl phenylboronic acid pinacol ester,4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-carbamic acid tert-butyl ester,pubchem5247,4-n-boc-aminomethyl phenylboronic acid,pinacol ester
3415442
tert-butyl N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CNC(=O)OC(C)(C)C
Melting Point 68.0°C to 69.0°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 333.24
Percent Purity 97%
Chemical Name or Material 4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning