Alfa Aesar™ 4-tert-Butyl-2,6-diaminoanisole, 97%

Manufacturer: Alfa Aesar™ H5587814

Quantity 25g

 View more versions of this product

Catalog No. AAH5587814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 473269-70-4
    Formula Weight 194.28
    MDL Number MFCD03701559
    Chemical Name or Material 4-tert-Butyl-2,6-diaminoanisole

    Specifications

    Formula Weight 194.28
    Melting Point 111° to 113°C
    Physical Form Powder
    MDL Number MFCD03701559
    CAS 473269-70-4
    Color White to orange
    Quantity 25g
    Chemical Name or Material 4-tert-Butyl-2,6-diaminoanisole
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit