Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(tert-Butyl)benzylbromide, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
18880-00-7
C11H15Br
227.14
MFCD00000180
QZNQSIHCDAGZIA-UHFFFAOYSA-N
4-tert-butylbenzyl bromide,p-tert-butylbenzyl bromide,1-bromomethyl-4-tert-butyl benzene,4-tert-butylbenzylbromide,1-bromomethyl-4-tert-butylbenzene,4-tert-butyl benzylbromide,p-t-butylbenzyl bromide,1-bromomethyl-4-1,1-dimethylethyl benzene,4-t-butylbenzylbromide,4-tert-buthylbenzyl bromide
87836
1-(bromomethyl)-4-tert-butylbenzene
CC(C)(C)C1=CC=C(C=C1)CBr
18880-00-7
C11H15Br
227.14
MFCD00000180
QZNQSIHCDAGZIA-UHFFFAOYSA-N
4-tert-butylbenzyl bromide,p-tert-butylbenzyl bromide,1-bromomethyl-4-tert-butyl benzene,4-tert-butylbenzylbromide,1-bromomethyl-4-tert-butylbenzene,4-tert-butyl benzylbromide,p-t-butylbenzyl bromide,1-bromomethyl-4-1,1-dimethylethyl benzene,4-t-butylbenzylbromide,4-tert-buthylbenzyl bromide
87836
1-(bromomethyl)-4-tert-butylbenzene
CC(C)(C)C1=CC=C(C=C1)CBr
Melting Point 8°C to 12°C
Boiling Point 90°C to 94°C (1.5mmHg)
Flash Point >109°C
Specific Gravity 1.236
Refractive Index 1.5445 to 1.5465
Density 1.2360g/mL
Color Yellow
Infrared Spectrum Authentic
Quantity 100g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (Argentometry)
Linear Formula (CH3)3CC6H4CH2Br
Formula Weight 227.14
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-(tert-Butyl)benzylbromide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 242-643-1