Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(tert-Butyl)-o-phenylenediamine, 97%

Manufacturer:  Alfa Aesar™ H32205MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH32205MD


  Check Availability
Add to cart

Specifications

Description

Applications
4-(tert-Butyl)-o-phenylenediamine, is used as a as a co-catalyst for the polymerization of o-phenylenediamine and its derivatives.

Solubility
Soluble in most common organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

68176-57-8
164.25
UN2811
4-(tert-Butyl)-o-phenylenediamine
C10H16N2
MFCD00052695
4-(tert-Butyl)-1,2-diaminobenzene

Specifications

68176-57-8
164.25
UN2811
98° to 100°C
4-(tert-Butyl)-o-phenylenediamine
C10H16N2
MFCD00052695
4-(tert-Butyl)-1,2-diaminobenzene
250mg
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit