Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-tert-Butylaniline, 99%

Manufacturer:  Alfa Aesar™ A1092522

Quantity 100g

 View more versions of this product

Catalog No. AAA1092522


  Check Availability
Add to cart

Specifications

Description

Applications
4-tert-Butylaniline reacts with formic acid to produce formic acid-(4-tert-butyl-anilide). This reaction could happen in the presence of the solvent of toluene in the condition of heating. 4-tert-Butylaniline is used in the synthesis of 4-tert-Butyl-4 ft.,4″-dinitrotriphenylamine, new triphenylamine-containing diamine monomer and 2-oxopyrimido[4,5-d]pyrimidin-5(6H)-one.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong acids, acid chlorides, acid anhydrides and chloroformates.
Description & Specifications

Chemical Identifiers

769-92-6
149.23
2205786
C10H15N
MFCD00007899
4-tert-Butylaniline

Specifications

769-92-6
149.23
2205786
15° to 16°C
100g
99%
C10H15N
MFCD00007899
235°C
101°C (213°F)
4-tert-Butylaniline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit