Alfa Aesar™ 4-tert-Butylanisole, 98%

Manufacturer: Alfa Aesar™ B2535722

Quantity 100g

 View more versions of this product

Catalog No. AAB2535722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5396-38-3
    Molecular Formula C11H16O
    Formula Weight 164.25
    MDL Number MFCD00026309
    Beilstein 2041952
    Synonym 4-Methoxy-tert-butylbenzene; 4-Methoxy-tert-butylbenzene
    Chemical Name or Material 4-tert-Butylanisole

    Specifications

    Formula Weight 164.25
    Molecular Formula C11H16O
    MDL Number MFCD00026309
    Synonym 4-Methoxy-tert-butylbenzene; 4-Methoxy-tert-butylbenzene
    Boiling Point 222°C
    Melting Point 18°C
    Flash Point 94°C (201°F)
    Quantity 100g
    TSCA TSCA
    CAS 5396-38-3
    Beilstein 2041952
    Chemical Name or Material 4-tert-Butylanisole
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit