Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-tert-Butylbenzenesulfonyl chloride, 98%

Click to view available options
Quantity:
10g
50g
15084-51-2
C10H13ClO2S
232.722
MFCD00007449
YEZADZMMVHWFIY-UHFFFAOYSA-N
4-t-butylbenzenesulfonyl chloride,4-tert-butyl benzene-1-sulfonyl chloride,4-tert-butylbenzene-1-sulfonyl chloride,4-tert-butylbenzenesulfonylchloride,4-tert-butyl benzenesulfonyl chloride,4-tert-butyl-benzenesulfonyl chloride,4-tert-butylphenylsulfonyl chloride,p-tert-butylbenzenesulfonyl chloride,4-tert butylbenzenesulfonyl chloride,4-tert-butyl phenylsulfonyl chloride
139882
4-tert-butylbenzenesulfonyl chloride
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl
15084-51-2
C10H13ClO2S
232.722
MFCD00007449
YEZADZMMVHWFIY-UHFFFAOYSA-N
4-t-butylbenzenesulfonyl chloride,4-tert-butyl benzene-1-sulfonyl chloride,4-tert-butylbenzene-1-sulfonyl chloride,4-tert-butylbenzenesulfonylchloride,4-tert-butyl benzenesulfonyl chloride,4-tert-butyl-benzenesulfonyl chloride,4-tert-butylphenylsulfonyl chloride,p-tert-butylbenzenesulfonyl chloride,4-tert butylbenzenesulfonyl chloride,4-tert-butyl phenylsulfonyl chloride
139882
4-tert-butylbenzenesulfonyl chloride
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl
Melting Point 80°C to 83°C
Odor Irritating
Quantity 50g
UN Number UN3261
Beilstein 1104346
Formula Weight 232.73
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzenesulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.