Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylbenzonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
4210-32-6
C11H13N
159.232
MFCD00075840
IIZURLNRIMKEDL-UHFFFAOYSA-N
4-tert-butyl benzonitrile,4-t-butylbenzonitrile,benzonitrile, 4-1,1-dimethylethyl,4-tert-butylcyanobenzene,4-tert-butyl-benzonitrile,p-t-butylbenzonitrile,acmc-209jnk,p-tert-butylbenzonitrile,benzonitrile, p-tert-butyl,ksc219k6d
77883
4-tert-butylbenzonitrile
CC(C)(C)C1=CC=C(C=C1)C#N
4210-32-6
C11H13N
159.232
MFCD00075840
IIZURLNRIMKEDL-UHFFFAOYSA-N
4-tert-butyl benzonitrile,4-t-butylbenzonitrile,benzonitrile, 4-1,1-dimethylethyl,4-tert-butylcyanobenzene,4-tert-butyl-benzonitrile,p-t-butylbenzonitrile,acmc-209jnk,p-tert-butylbenzonitrile,benzonitrile, p-tert-butyl,ksc219k6d
77883
4-tert-butylbenzonitrile
CC(C)(C)C1=CC=C(C=C1)C#N
Color Colorless
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5165 to 1.5185 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 159.23
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4-tert-Butylbenzonitrile, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning