Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-tert-Butyldimethylsiloxy-1-butanol, 97%

Manufacturer:  Alfa Aesar™ H5528506

Quantity 5g

 View more versions of this product

Catalog No. AAH5528506


  Check Availability
Add to cart

Specifications

Description

Solubility
Slightly Soluble in water.(2.0 g/L) (25°C). Soluble in Dichloromethane, Ethyl Acetate, Methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
Description & Specifications

Chemical Identifiers

87184-99-4
(CH3)3CSi(CH3)2O(CH2)4OH
MFCD04974070
C10H24O2Si
204.38
4-tert-Butyldimethylsiloxy-1-butanol

Specifications

87184-99-4
(CH3)3CSi(CH3)2O(CH2)4OH
MFCD04974070
5g
4-tert-Butyldimethylsiloxy-1-butanol
C10H24O2Si
204.38
63° to 66°C (0.3mmHg)
105°C (221°F)
97%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit