Alfa Aesar™ 4-tert-Butyldimethylsiloxy-1-butanol, 97%

Manufacturer: Alfa Aesar™ H5528514

Quantity 25g

 View more versions of this product

Catalog No. AAH5528514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87184-99-4
    Molecular Formula C10H24O2Si
    Linear Formula (CH3)3CSi(CH3)2O(CH2)4OH
    Formula Weight 204.38
    MDL Number MFCD04974070
    Chemical Name or Material 4-tert-Butyldimethylsiloxy-1-butanol

    Specifications

    Formula Weight 204.38
    Boiling Point 63° to 66°C (0.3mmHg)
    Quantity 25g
    Flash Point 105°C (221°F)
    Molecular Formula C10H24O2Si
    Linear Formula (CH3)3CSi(CH3)2O(CH2)4OH
    MDL Number MFCD04974070
    CAS 87184-99-4
    Chemical Name or Material 4-tert-Butyldimethylsiloxy-1-butanol
    Assay 97%

    Description

    Solubility
    Slightly Soluble in water.(2.0 g/L) (25°C). Soluble in Dichloromethane, Ethyl Acetate, Methanol

    Notes
    Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit