Alfa Aesar™ 4-tert-Butylphenyl glycidyl ether, 95%

Manufacturer: Alfa Aesar™ H5594514

Quantity 25g

 View more versions of this product

Catalog No. AAH5594514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3101-60-8
    Molecular Formula C13H18O2
    Formula Weight 206.28
    MDL Number MFCD00005136
    Chemical Name or Material 4-tert-Butylphenyl glycidyl ether

    Specifications

    Formula Weight 206.28
    Boiling Point 165° to 170°C (14mmHg)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P362-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 1.038g/mL
    Physical Form Liquid
    Refractive Index 1.5150
    Color Colorless
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H317
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    TSCA TSCA
    MDL Number MFCD00005136
    Flash Point 102°C (216°F)
    Quantity 25g
    RTECS Number TX4250000
    Molecular Formula C13H18O2
    CAS 3101-60-8
    Chemical Name or Material 4-tert-Butylphenyl glycidyl ether
    Assay 95%

    Description

    Applications
    4-tert-Butylphenyl glycidyl ether is used in paint industry and serves as organic coating in Paint, boat Paint, building coating material. It is also used as an epoxy reactive diluent.

    Solubility
    Slightly soluble in water.

    Notes
    Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H317
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P305+P351+P338-P362-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit