Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylphenylacetic acid 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
32857-63-9
C12H16O2
192.258
MFCD00082593
RUAYXHSDAMWEDR-UHFFFAOYSA-N
4-tert-butylphenylacetic acid,4-tert-butylphenyl acetic acid,2-4-tert-butylphenyl acetic acid,4-t-butylphenylacetic acid,2-4-tert-butyl phenyl acetic acid,benzeneacetic acid, 4-1,1-dimethylethyl,4-tert-butyl-phenyl-acetic acid,tb2,acmc-1cq6j,asischem y85510
118343
2-(4-tert-butylphenyl)acetic acid
CC(C)(C)C1=CC=C(C=C1)CC(=O)O
32857-63-9
C12H16O2
192.258
MFCD00082593
RUAYXHSDAMWEDR-UHFFFAOYSA-N
4-tert-butylphenylacetic acid,4-tert-butylphenyl acetic acid,2-4-tert-butylphenyl acetic acid,4-t-butylphenylacetic acid,2-4-tert-butyl phenyl acetic acid,benzeneacetic acid, 4-1,1-dimethylethyl,4-tert-butyl-phenyl-acetic acid,tb2,acmc-1cq6j,asischem y85510
118343
2-(4-tert-butylphenyl)acetic acid
CC(C)(C)C1=CC=C(C=C1)CC(=O)O
Quantity 1g
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 72°C to 82°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3)3CC6H4CH2COOH
Formula Weight 192.26
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-tert-Butylphenylacetic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.