Alfa Aesar™ 4-tert-Butylphenylacetylene, 90+%

Manufacturer: Alfa Aesar™ 4392018

Quantity 50g

 View more versions of this product

Catalog No. AA4392018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 772-38-3
    Molecular Formula C12H14
    Formula Weight 158.24
    MDL Number MFCD00190197
    UN Number UN3082
    Chemical Name or Material 4-tert-Butylphenylacetylene

    Specifications

    Formula Weight 158.24
    Health Hazard 3 GHS P Statement
    P273-P501a
    Avoid release to the environment.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 0.878g/mL
    CAS 772-38-3
    Refractive Index 1.5297
    Molecular Formula C12H14
    MDL Number MFCD00190197
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Boiling Point 130°C (40mmHg)
    Odor Characteristic
    Physical Form Liquid
    UN Number UN3082
    Flash Point 82°C (180°F)
    Quantity 50g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Chemical Name or Material 4-tert-Butylphenylacetylene
    Assay ≥90%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P273-P501a
    Avoid release to the environment.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit