Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylphenylacetylene, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
772-38-3
C12H14
158.24
MFCD00190197
ZSYQVVKVKBVHIL-UHFFFAOYSA-N
1-tert-butyl-4-ethynylbenzene,4-tert-butyl phenylacetylene,4-tert-butylphenylacetylene,benzene, 1-1,1-dimethylethyl-4-ethynyl,acmc-20amtx,4-t-butylphenylacetylene,4-tert-butylphenyl ethyne,4-tert.butylphenylacetylen,4-tert-butyl phenylethyne,4-tert-butylphenyl acetylene
2757302
1-tert-butyl-4-ethynylbenzene
CC(C)(C)C1=CC=C(C=C1)C#C
772-38-3
C12H14
158.24
MFCD00190197
ZSYQVVKVKBVHIL-UHFFFAOYSA-N
1-tert-butyl-4-ethynylbenzene,4-tert-butyl phenylacetylene,4-tert-butylphenylacetylene,benzene, 1-1,1-dimethylethyl-4-ethynyl,acmc-20amtx,4-t-butylphenylacetylene,4-tert-butylphenyl ethyne,4-tert.butylphenylacetylen,4-tert-butyl phenylethyne,4-tert-butylphenyl acetylene
2757302
1-tert-butyl-4-ethynylbenzene
CC(C)(C)C1=CC=C(C=C1)C#C
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 130.0°C (40.0 mmHg)
Color Brown-Khaki to Green
Quantity 5g
Refractive Index 1.5280 to 1.5300
Flash Point 82.2°C
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula CH≡CC6H4C(CH3)3
Formula Weight 158.24
Physical Form Powder and/or Chunks
Percent Purity 96%
Chemical Name or Material 4-tert-Butylphenylacetylene
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning