Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
123324-71-0
C10H15BO2
178.04
MNJYZNVROSZZQC-UHFFFAOYSA-N
4-t-butylphenylboronic acid,4-tert-butylbenzeneboronic acid,4-tert-butyl phenyl boronic acid,4-tert-butylphenyl boronic acid,4-tert-butyl phenylboronic acid,4-tert-butyl-phenyl boronic acid,4-tert-butylphenyl boranediol,p-t-butylphenylboronic acid,4-tertbutylphenylboronic acid
2734320
(4-tert-butylphenyl)boronic acid
B(C1=CC=C(C=C1)C(C)(C)C)(O)O
123324-71-0
C10H15BO2
178.04
MNJYZNVROSZZQC-UHFFFAOYSA-N
4-t-butylphenylboronic acid,4-tert-butylbenzeneboronic acid,4-tert-butyl phenyl boronic acid,4-tert-butylphenyl boronic acid,4-tert-butyl phenylboronic acid,4-tert-butyl-phenyl boronic acid,4-tert-butylphenyl boranediol,p-t-butylphenylboronic acid,4-tertbutylphenylboronic acid
2734320
(4-tert-butylphenyl)boronic acid
B(C1=CC=C(C=C1)C(C)(C)C)(O)O
Color White
Quantity 25g
Melting Point 191.0°C to 196.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Linear Formula (CH3)3CC6H4B(OH)2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,thf,dmso,dmf,methanol
Formula Weight 178.04
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-tert-Butylphenylboronic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning