Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-tert-Butylthiophenol gold nanoparticles, 2% (w/v) soln. in toluene, 2-5nm particles

Manufacturer:  Alfa Aesar™ H28235AA

Quantity 5mL

 View more versions of this product

Catalog No. AAH28235AA


  Check Availability
Add to cart

Specifications

Description

Applications
The study of gold nanoparticles is very captivating because of their significance and applications as catalysts in restructuring technologies that are used for manufacturing, medicine, energy production, transportation, computation, communication, and environmental changes.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

MFCD09881474
UN1294
Au-tert-butylbenzenethiol
4-tert-Butylthiophenol gold nanoparticles, 2 to 5nm particles

Specifications

MFCD09881474
Au-tert-butylbenzenethiol
4°C (39°F)
2% (w/v) soln. in toluene
4-tert-Butylthiophenol gold nanoparticles, 2 to 5nm particles
UN1294
111°C
Aromatic
5mL

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE

Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit