Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Thien-3-yl)benzoic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
29886-64-4
C11H8O2S
204.25
FISAUHGRILVMDP-UHFFFAOYSA-N
4-3-thienyl benzoic acid,4-thiophen-3-yl benzoic acid,4-thiophen-3-yl-benzoic acid,benzoic acid, 4-3-thienyl,4-thien-3-ylbenzoic acid,4-3-thienyl-benzoic acid,maybridge3_004545,acmc-1cisg
736862
4-thiophen-3-ylbenzoic acid
C1=CC(=CC=C1C2=CSC=C2)C(=O)O
29886-64-4
C11H8O2S
204.25
FISAUHGRILVMDP-UHFFFAOYSA-N
4-3-thienyl benzoic acid,4-thiophen-3-yl benzoic acid,4-thiophen-3-yl-benzoic acid,benzoic acid, 4-3-thienyl,4-thien-3-ylbenzoic acid,4-3-thienyl-benzoic acid,maybridge3_004545,acmc-1cisg
736862
4-thiophen-3-ylbenzoic acid
C1=CC(=CC=C1C2=CSC=C2)C(=O)O
Packaging Glass Bottle
Melting Point 281.0°C to 283.0°C
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Formula Weight 204.25
Percent Purity 97%
Chemical Name or Material 4-(Thien-3-yl)benzoic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

Warning