Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Thiouracil, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
591-28-6
C4H4N2OS
128.15
MFCD00090842
OVONXEQGWXGFJD-UHFFFAOYSA-N
4-thiouracil,2-hydroxy-4 1h-pyrimidinethione,4-thioxo-3,4-dihydropyrimidin-2 1h-one,2-hydroxy-4-mercaptopyrimidine,uracil, 4-thio,4-mercaptopyrimidin-2-ol,4-sulfanylpyrimidin-2-ol,2-hydroxy-4-thiopyrimidine,2 1h-pyrimidinone, 3,4-dihydro-4-thioxo,2 1h-pyrimidinone, 4-mercapto-9ci
2734394
4-sulfanylidene-1H-pyrimidin-2-one
C1=CNC(=O)NC1=S
591-28-6
C4H4N2OS
128.15
MFCD00090842
OVONXEQGWXGFJD-UHFFFAOYSA-N
4-thiouracil,2-hydroxy-4 1h-pyrimidinethione,4-thioxo-3,4-dihydropyrimidin-2 1h-one,2-hydroxy-4-mercaptopyrimidine,uracil, 4-thio,4-mercaptopyrimidin-2-ol,4-sulfanylpyrimidin-2-ol,2-hydroxy-4-thiopyrimidine,2 1h-pyrimidinone, 3,4-dihydro-4-thioxo,2 1h-pyrimidinone, 4-mercapto-9ci
2734394
4-sulfanylidene-1H-pyrimidin-2-one
C1=CNC(=O)NC1=S
Melting Point 295°C
Infrared Spectrum Authentic
Appearance (5% sol. in 1N NaOH) : clear and faint yellow
Quantity 250mg
Packaging Glass bottle
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 128.15
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Thiouracil, 97%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.

GHS Signal Word: Warning