Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Tolylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
5720-05-8
C7H9BO2
135.96
MFCD00039138
BIWQNIMLAISTBV-UHFFFAOYSA-N
4-tolylboronic acid,p-tolylboronic acid,4-methylphenyl boronic acid,p-tolueneboronic acid,4-methylbenzeneboronic acid,p-methylbenzeneboronic acid,boronic acid, 4-methylphenyl,boronic acid, p-tolyl,4-methylphenylboric acid,tolyl boronic acid
79799
5720-05-8
C7H9BO2
135.96
MFCD00039138
BIWQNIMLAISTBV-UHFFFAOYSA-N
4-tolylboronic acid,p-tolylboronic acid,4-methylphenyl boronic acid,p-tolueneboronic acid,4-methylbenzeneboronic acid,p-methylbenzeneboronic acid,boronic acid, 4-methylphenyl,boronic acid, p-tolyl,4-methylphenylboric acid,tolyl boronic acid
79799
Melting Point 256°C to 263°C
Color White
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C6H4B(OH)2
Formula Weight 135.96
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Tolylboronic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physicia

GHS Signal Word: Warning