Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-trans-Hydroxy-L-proline ethyl ester hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
100g
33996-30-4
C7H14ClNO3
195.643
MFCD00040589
HHXSZDXMSRXWJV-IBTYICNHSA-N
h-hyp-oet hcl,h-hyp-oet.hcl,2s,4r-ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride,l-4-hydroxyproline ethyl ester hydrochloride,ethyl trans-4-hydroxy-l-prolinate hydrochloride,trans-4-hydroxy-l-proline ethyl ester hydrochloride,ethyl 2s,4r-4-hydroxypyrrolidine-2-carboxylate hydrochloride,ethyl trans-4-hydroxy-l-prolinate hcl,2s,4r---ethyl 4-hydroxy-2-pyrrolidincarboxylate hydrochloride 99+%,2s,4r-4-hydroxyproline ethyl ester hydrochloride
12709304
ethyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride
CCOC(=O)C1CC(CN1)O.Cl
33996-30-4
C7H14ClNO3
195.643
MFCD00040589
HHXSZDXMSRXWJV-IBTYICNHSA-N
h-hyp-oet hcl,h-hyp-oet.hcl,2s,4r-ethyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride,l-4-hydroxyproline ethyl ester hydrochloride,ethyl trans-4-hydroxy-l-prolinate hydrochloride,trans-4-hydroxy-l-proline ethyl ester hydrochloride,ethyl 2s,4r-4-hydroxypyrrolidine-2-carboxylate hydrochloride,ethyl trans-4-hydroxy-l-prolinate hcl,2s,4r---ethyl 4-hydroxy-2-pyrrolidincarboxylate hydrochloride 99+%,2s,4r-4-hydroxyproline ethyl ester hydrochloride
12709304
ethyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride
CCOC(=O)C1CC(CN1)O.Cl
Beilstein 3656870
Formula Weight 195.64
Physical Form Powder
Percent Purity 97%
Color White
Quantity 100g
Chemical Name or Material 4-trans-Hydroxy-L-proline ethyl ester hydrochloride

storageNote1 : 2° to 8°C