Alfa Aesar™ 4-Tridecanone, 98%

Manufacturer: Alfa Aesar™ B2472106

Quantity 5g

 View more versions of this product

Catalog No. AAB2472106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 26215-90-7
    Molecular Formula CH3(CH2)8COCH2CH2CH3
    Formula Weight 198.35
    MDL Number MFCD00027351
    Synonym n-Propyl n-nonyl ketone
    Chemical Name or Material 4-Tridecanone

    Specifications

    Formula Weight 198.35
    Molecular Formula CH3(CH2)8COCH2CH2CH3
    MDL Number MFCD00027351
    CAS 26215-90-7
    Synonym n-Propyl n-nonyl ketone
    Melting Point 18° to 19°C
    Quantity 5g
    Chemical Name or Material 4-Tridecanone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit