Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Tridecanone, 98%

Click to view available options
Quantity:
1g
5g
25g
26215-90-7
C13H26O
198.35
MFCD00027351
JWSRUPAFLPWLNO-UHFFFAOYSA-N
4-tridecanone,n-nonyl n-propyl ketone,tridecane-4-one,acmc-1chwt,n-propyl n-nonyl ketone
98673
tridecan-4-one
CCCCCCCCCC(=O)CCC
26215-90-7
C13H26O
198.35
MFCD00027351
JWSRUPAFLPWLNO-UHFFFAOYSA-N
4-tridecanone,n-nonyl n-propyl ketone,tridecane-4-one,acmc-1chwt,n-propyl n-nonyl ketone
98673
tridecan-4-one
CCCCCCCCCC(=O)CCC
Quantity 25g
Melting Point 18°C to 19°C
Formula Weight 198.35
Percent Purity 98%
Chemical Name or Material 4-Tridecanone