Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)aniline 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
250g
461-82-5
C7H6F3NO
177.126
MFCD00041314
XUJFOSLZQITUOI-UHFFFAOYSA-N
4-trifluoromethoxy aniline,4-trifluoromethoxy-phenylamine,4-trifluoromethoxy benzenamine,p-trifluoromethoxyaniline,4-trifluoroethoxy aniline,benzenamine, 4-trifluoromethoxy,unii-p40l42cvf6,alpha,alpha,alpha-trifluoro-p-anisidine,p-aminotrifluoromethoxybenzene,4-amino-alpha,alpha,alpha-trifluoroanisole
600848
4-(trifluoromethoxy)aniline
C1=CC(=CC=C1N)OC(F)(F)F
461-82-5
C7H6F3NO
177.126
MFCD00041314
XUJFOSLZQITUOI-UHFFFAOYSA-N
4-trifluoromethoxy aniline,4-trifluoromethoxy-phenylamine,4-trifluoromethoxy benzenamine,p-trifluoromethoxyaniline,4-trifluoroethoxy aniline,benzenamine, 4-trifluoromethoxy,unii-p40l42cvf6,alpha,alpha,alpha-trifluoro-p-anisidine,p-aminotrifluoromethoxybenzene,4-amino-alpha,alpha,alpha-trifluoroanisole
600848
4-(trifluoromethoxy)aniline
C1=CC(=CC=C1N)OC(F)(F)F
Quantity 5g
Color Undesignated
Boiling Point 83°C
UN Number 2810
Formula Weight 177.13
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 4-(Trifluoromethoxy)aniline