Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Trifluoromethoxy)benzoyl chloride, 97%

Click to view available options
Quantity:
1g
5g
25g
36823-88-8
C8H4ClF3O2
224.563
MFCD00052329
ZXKKOFJYPRJFIE-UHFFFAOYSA-N
4-trifluoromethoxy benzoyl chloride,p-trifluoromethoxybenzoyl chloride,benzoyl chloride, 4-trifluoromethoxy,4-trifluoromethoxy benzene-1-carbonyl chloride,4-trifluoromethoxy benzoylchloride,4-trifluoromethoxy-benzoyl chloride,p-trifluoromethoxy benzoyl chloride,acmc-209ip1,ksc495i1f,p-trifluoromethoxybenzoylchloride
142110
4-(trifluoromethoxy)benzoyl chloride
C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F
36823-88-8
C8H4ClF3O2
224.563
MFCD00052329
ZXKKOFJYPRJFIE-UHFFFAOYSA-N
4-trifluoromethoxy benzoyl chloride,p-trifluoromethoxybenzoyl chloride,benzoyl chloride, 4-trifluoromethoxy,4-trifluoromethoxy benzene-1-carbonyl chloride,4-trifluoromethoxy benzoylchloride,4-trifluoromethoxy-benzoyl chloride,p-trifluoromethoxy benzoyl chloride,acmc-209ip1,ksc495i1f,p-trifluoromethoxybenzoylchloride
142110
4-(trifluoromethoxy)benzoyl chloride
C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F
Density 1.425
Sensitivity Moisture sensitive
Refractive Index 1.472
Boiling Point 55°C to 56°C (2mmHg)
Quantity 1g
Flash Point 90°C (194°F)
UN Number UN3265
Beilstein 2723543
Formula Weight 224.57
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)benzoyl chloride
Show More Show Less

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures