Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)phenylacetonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
49561-96-8
C9H6F3NO
201.15
MFCD00061240
LYFCAROXYJTUQF-UHFFFAOYSA-N
4-trifluoromethoxy phenylacetonitrile,2-4-trifluoromethoxy phenyl acetonitrile,4-trifluoromethoxybenzyl cyanide,4-trifluoromethoxy benzyl cyanide,benzeneacetonitrile, 4-trifluoromethoxy,4-trifluoromethoxy-phenyl-acetonitrile,4-trifluoromethoxy phenyl acetonitrile,4-trifluoromethoxybenzylcyanide,1-4-trifluoromethoxy phenyl acetonitrile
142676
2-[4-(trifluoromethoxy)phenyl]acetonitrile
C1=CC(=CC=C1CC#N)OC(F)(F)F
49561-96-8
C9H6F3NO
201.15
MFCD00061240
LYFCAROXYJTUQF-UHFFFAOYSA-N
4-trifluoromethoxy phenylacetonitrile,2-4-trifluoromethoxy phenyl acetonitrile,4-trifluoromethoxybenzyl cyanide,4-trifluoromethoxy benzyl cyanide,benzeneacetonitrile, 4-trifluoromethoxy,4-trifluoromethoxy-phenyl-acetonitrile,4-trifluoromethoxy phenyl acetonitrile,4-trifluoromethoxybenzylcyanide,1-4-trifluoromethoxy phenyl acetonitrile
142676
2-[4-(trifluoromethoxy)phenyl]acetonitrile
C1=CC(=CC=C1CC#N)OC(F)(F)F
Quantity 1g
Color Yellow
Refractive Index 1.4480 to 1.45
Melting Point 28°C to 30°C
Flash Point 113°C
Specific Gravity 1.28
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.2800g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula F3COC6H4CH2CN
Formula Weight 201.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)phenylacetonitrile
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning