Alfa Aesar™ 4-Trifluoromethyl-1H-pyrazole, 97%

Manufacturer: Alfa Aesar™ H34396MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH34396MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 52222-73-8
    Molecular Formula C4H3F3N2
    Formula Weight 136.1
    MDL Number MFCD11226572
    Chemical Name or Material 4-Trifluoromethyl-1H-pyrazole

    Specifications

    Formula Weight 136.1
    Molecular Formula C4H3F3N2
    MDL Number MFCD11226572
    CAS 52222-73-8
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 76° to 78°C
    Quantity 250mg
    Chemical Name or Material 4-Trifluoromethyl-1H-pyrazole
    Assay 97%

    Description

    Applications
    It is used as pharmaceutical intermediates. The palladium-catalyzed cross-coupling reactions of 5-tributylstannyl-4-fluoro-1H-pyrazole with aryl iodides provided high yields of the corresponding 5-aryl-4-fluoro-1H-pyrazoles.

    Notes
    Incompatible with strong oxidizing agents. Store at room temperature.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit