Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Matrix Scientific 4'-(Trifluoromethyl)acetophenone, 709-63-7, MFCD00000401, 25g
Manufacturer: Matrix Scientific 00649625G
Molecular Formula C9H7F3O, Purity 98%, Molecular Weight 188.15, Melting Point 30-33°, Boiling Point 79-80/8mm
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title