Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-(Trifluoromethyl)acetophenone, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
709-63-7
C9H7F3O
188.15
MFCD00000401
HHAISVSEJFEWBZ-UHFFFAOYSA-N
4'-trifluoromethyl acetophenone,1-4-trifluoromethyl phenyl ethanone,1-4-trifluoromethyl phenyl ethan-1-one,4-trifluoromethylacetophenone,4-acetylbenzotrifluoride,4-trifluoromethyl acetophenone,p-trifluoromethylacetophenone,4'-trifluoromethylacetophenone,ethanone, 1-4-trifluoromethyl phenyl
69731
1-[4-(trifluoromethyl)phenyl]ethanone
CC(=O)C1=CC=C(C=C1)C(F)(F)F
709-63-7
C9H7F3O
188.15
MFCD00000401
HHAISVSEJFEWBZ-UHFFFAOYSA-N
4'-trifluoromethyl acetophenone,1-4-trifluoromethyl phenyl ethanone,1-4-trifluoromethyl phenyl ethan-1-one,4-trifluoromethylacetophenone,4-acetylbenzotrifluoride,4-trifluoromethyl acetophenone,p-trifluoromethylacetophenone,4'-trifluoromethylacetophenone,ethanone, 1-4-trifluoromethyl phenyl
69731
1-[4-(trifluoromethyl)phenyl]ethanone
CC(=O)C1=CC=C(C=C1)C(F)(F)F
Refractive Index 1.4604
Boiling Point 79°C to 80°C (8.0mmHg)
Flash Point 84°C
Density 0.9200g/mL
Specific Gravity 0.92
Color White to Yellow
Packaging Glass bottle
Quantity 1g
Melting Point 30°C to 33°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula CF3C6H4COCH3
Formula Weight 188.15
Physical Form Low Melting Solid
Percent Purity 99%
Chemical Name or Material 4'-(Trifluoromethyl)acetophenone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-913-