Learn More
4-(Trifluoromethyl)benzeneboronic acid, 98%
Description
4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-(Trifluoromethyl)benzeneboronic acid is used of this drug is to N-arylate imidazoles and amines with copper-exchanged fluorapatite, as well as used in microwave-promoted cross-coupling with acid chlorides leading to aryl ketones.
Solubility
Soluble in DMSO, not in water
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 238°C to 242°C |
| Quantity | 25 g |
| Beilstein | 3544189 |
| Solubility Information | Soluble in DMSO,not in water |
| Formula Weight | 189.93 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-(Trifluoromethyl)benzeneboronic acid |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.