Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Trifluoromethyl)benzenesulfonamide, 97%

Click to view available options
Quantity:
1g
5g
25g
830-43-3
C7H6F3NO2S
225.185
MFCD00159251
TVHXQQJDMHKGGK-UHFFFAOYSA-N
4-trifluoromethyl benzenesulfonamide,4-trifluoromethyl benzene-1-sulfonamide,benzenesulfonamide, p-trifluoromethyl,4-trifluoromethylbenzenesulphonamide,p-trifluoromethyl benzenesulfonamide,4-trifluoromethyl benzenesulphonamide,benzenesulfonamide, 4-trifluoromethyl,p-trifluoromethylbenzenesulfonamide,alpha,alpha,alpha-trifluoro-p-toluenesulphonamide,pubchem11767
70018
4-(trifluoromethyl)benzenesulfonamide
C1=CC(=CC=C1C(F)(F)F)S(=O)(=O)N
830-43-3
C7H6F3NO2S
225.185
MFCD00159251
TVHXQQJDMHKGGK-UHFFFAOYSA-N
4-trifluoromethyl benzenesulfonamide,4-trifluoromethyl benzene-1-sulfonamide,benzenesulfonamide, p-trifluoromethyl,4-trifluoromethylbenzenesulphonamide,p-trifluoromethyl benzenesulfonamide,4-trifluoromethyl benzenesulphonamide,benzenesulfonamide, 4-trifluoromethyl,p-trifluoromethylbenzenesulfonamide,alpha,alpha,alpha-trifluoro-p-toluenesulphonamide,pubchem11767
70018
4-(trifluoromethyl)benzenesulfonamide
C1=CC(=CC=C1C(F)(F)F)S(=O)(=O)N
Quantity 25g
Melting Point 178°C to 182°C
Beilstein 2695323
Formula Weight 225.19
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)benzenesulfonamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DB3200000