Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)benzophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
728-86-9
C14H9F3O
250.22
MFCD00000400
OHTYZZYAMUVKQS-UHFFFAOYSA-N
4-trifluoromethyl benzophenone, 4-trifluoromethylbenzophenone, phenyl 4-trifluoromethyl phenyl methanone, p-trifluoromethylbenzophenone, phenyl-4-trifluoromethyl phenyl methanone, methanone, phenyl 4-trifluoromethyl phenyl, fxffr dvr, phenyl 4-trifluoromethyl phenyl ketone, acmc-20amto
69767
phenyl-[4-(trifluoromethyl)phenyl]methanone
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(F)(F)F
728-86-9
C14H9F3O
250.22
MFCD00000400
OHTYZZYAMUVKQS-UHFFFAOYSA-N
4-trifluoromethyl benzophenone, 4-trifluoromethylbenzophenone, phenyl 4-trifluoromethyl phenyl methanone, p-trifluoromethylbenzophenone, phenyl-4-trifluoromethyl phenyl methanone, methanone, phenyl 4-trifluoromethyl phenyl, fxffr dvr, phenyl 4-trifluoromethyl phenyl ketone, acmc-20amto
69767
phenyl-[4-(trifluoromethyl)phenyl]methanone
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(F)(F)F
Quantity 5g
Infrared Spectrum Authentic
Color White to Beige
Melting Point 113°C to 116°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CF3C6H4COC6H5
Beilstein 07, IV, 1404
Formula Weight 250.22
Physical Form Crystalline Solid
Percent Purity ≥96.0%
Chemical Name or Material 4-(Trifluoromethyl)benzophenone, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-974-3