Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)benzyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
349-95-1
C8H7F3O
176.14
MFCD00004661
MOOUWXDQAUXZRG-UHFFFAOYSA-N
4-trifluoromethyl benzyl alcohol,4-trifluoromethyl phenyl methanol,p-trifluoromethylbenzyl alcohol,4-trifluoromethyl benzene methanol,4-trifluoromethyl benzylic alcohol,benzenemethanol, 4-trifluoromethyl,4-trifluoromethylbenzyl alcohol,ccris 5117,4-trifluoromethyl benzylalcohol
67684
[4-(trifluoromethyl)phenyl]methanol
C1=CC(=CC=C1CO)C(F)(F)F
349-95-1
C8H7F3O
176.14
MFCD00004661
MOOUWXDQAUXZRG-UHFFFAOYSA-N
4-trifluoromethyl benzyl alcohol,4-trifluoromethyl phenyl methanol,p-trifluoromethylbenzyl alcohol,4-trifluoromethyl benzene methanol,4-trifluoromethyl benzylic alcohol,benzenemethanol, 4-trifluoromethyl,4-trifluoromethylbenzyl alcohol,ccris 5117,4-trifluoromethyl benzylalcohol
67684
[4-(trifluoromethyl)phenyl]methanol
C1=CC(=CC=C1CO)C(F)(F)F
Refractive Index 1.4590 to 1.4610
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Melting Point 18.0°C to 20.0°C
Density 1.2800g/mL
Boiling Point 78.0°C to 80.0°C (4.0mmHg)
Color Colorless to Brown
Specific Gravity 1.28
Flash Point 100°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula CF3C6H4CH2OH
Formula Weight 176.14
Physical Form Liquid After Melting
Percent Purity 99%
Chemical Name or Material 4-(Trifluoromethyl)benzyl alcohol, 99%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

EINECSNumber : 206-494-6