Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-(Trifluoromethyl)cyclohexan-1-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Linear Formula | CF3C6H9(=O) |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 166.14 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Trifluoromethyl)cyclohexan-1-one |