Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Trifluoromethyl)diphenyl ether, 97%

Click to view available options
Quantity:
1g
5g
2367-02-4
C13H9F3O
238.21
MFCD01631641
UZJUDUZMPNCXPF-UHFFFAOYSA-N
4-trifluoromethyl diphenyl ether,1-phenoxy-4-trifluoromethyl benzene,4-phenoxy-1-trifluoromethyl benzene,4-phenoxybenzotrifluoride,trifluoromethyldiphenyl ether,4-trifluoromethyl diphenylether,4-phenoxy-alpha,alpha,alpha-trifluorotoluene
2777517
1-phenoxy-4-(trifluoromethyl)benzene
C1=CC=C(C=C1)OC2=CC=C(C=C2)C(F)(F)F
2367-02-4
C13H9F3O
238.21
MFCD01631641
UZJUDUZMPNCXPF-UHFFFAOYSA-N
4-trifluoromethyl diphenyl ether,1-phenoxy-4-trifluoromethyl benzene,4-phenoxy-1-trifluoromethyl benzene,4-phenoxybenzotrifluoride,trifluoromethyldiphenyl ether,4-trifluoromethyl diphenylether,4-phenoxy-alpha,alpha,alpha-trifluorotoluene
2777517
1-phenoxy-4-(trifluoromethyl)benzene
C1=CC=C(C=C1)OC2=CC=C(C=C2)C(F)(F)F
Applications
4-(Trifluoromethyl)diphenyl ether is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Storage temperature is 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.
Quantity 5g
Boiling Point 180°C (10mmHg)
Beilstein 1974851
Formula Weight 238.21
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)diphenyl ether

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning