Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Trifluoromethyl)indole, 97%

Click to view available options
Quantity:
250mg
1g
128562-95-8
C9H6F3N
185.149
MFCD09954766
YTVBZSLUNRYKID-UHFFFAOYSA-N
4-trifluoromethyl-1h-indole,4-trifluoromethyl indole,4-trifluoromethyl-indole,4-trifluoromethylindole,1h-indole, 4-trifluoromethyl
19803703
4-(trifluoromethyl)-1H-indole
C1=CC(=C2C=CNC2=C1)C(F)(F)F
128562-95-8
C9H6F3N
185.149
MFCD09954766
YTVBZSLUNRYKID-UHFFFAOYSA-N
4-trifluoromethyl-1h-indole,4-trifluoromethyl indole,4-trifluoromethyl-indole,4-trifluoromethylindole,1h-indole, 4-trifluoromethyl
19803703
4-(trifluoromethyl)-1H-indole
C1=CC(=C2C=CNC2=C1)C(F)(F)F
Quantity 1g
Beilstein 18343909
Formula Weight 185.15
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)indole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning