Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)-o-phenylenediamine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
368-71-8
C7H7F3N2
176.142
MFCD00042456
RQWJHUJJBYMJMN-UHFFFAOYSA-N
3,4-diaminobenzotrifluoride,4-trifluoromethyl benzene-1,2-diamine,4-trifluoromethyl-1,2-phenylenediamine,4-trifluoromethyl-o-phenylenediamine,1,2-benzenediamine, 4-trifluoromethyl,4-trifluoromethyl-1,2-benzenediamine,3,4-diamino-benzotrifluoride,3,4-diamino benzotrifluoride
520822
4-(trifluoromethyl)benzene-1,2-diamine
C1=CC(=C(C=C1C(F)(F)F)N)N
368-71-8
C7H7F3N2
176.142
MFCD00042456
RQWJHUJJBYMJMN-UHFFFAOYSA-N
3,4-diaminobenzotrifluoride,4-trifluoromethyl benzene-1,2-diamine,4-trifluoromethyl-1,2-phenylenediamine,4-trifluoromethyl-o-phenylenediamine,1,2-benzenediamine, 4-trifluoromethyl,4-trifluoromethyl-1,2-benzenediamine,3,4-diamino-benzotrifluoride,3,4-diamino benzotrifluoride
520822
4-(trifluoromethyl)benzene-1,2-diamine
C1=CC(=C(C=C1C(F)(F)F)N)N
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown-Gray
Melting Point 59°C to 62°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 176.14
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 4-(Trifluoromethyl)-o-phenylenediamine
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning