Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Trifluoromethyl)piperidine, 97%

Click to view available options
Quantity:
250mg
1g
5g
657-36-3
C6H10F3N
153.148
MFCD00102068
RDRQUUWCJTYHCT-UHFFFAOYSA-N
4-trifluoromethyl piperidine,4-trifluoromethyl-piperidine,piperidine, 4-trifluoromethyl,pubchem22645,4-trifluormethyl-piperidine,4-trifluoromethyl piperidine
136469
4-(trifluoromethyl)piperidine
C1CNCCC1C(F)(F)F
657-36-3
C6H10F3N
153.148
MFCD00102068
RDRQUUWCJTYHCT-UHFFFAOYSA-N
4-trifluoromethyl piperidine,4-trifluoromethyl-piperidine,piperidine, 4-trifluoromethyl,pubchem22645,4-trifluormethyl-piperidine,4-trifluoromethyl piperidine
136469
4-(trifluoromethyl)piperidine
C1CNCCC1C(F)(F)F
Color Colorless
Quantity 250mg
Formula Weight 153.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)piperidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning