Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)thiobenzamide, 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
72505-21-6
C8H6F3NS
205.2
IPRFNMJROWWFBH-UHFFFAOYSA-N
4-trifluoromethyl thiobenzamide,4-trifluoromethyl benzenecarbothioamide,4-trifluoromethylthiobenzamide,4-trifluoromethyl benzene-1-carbothioamide,4-trifluoromethyl benzothioamide,4-trifluoromethyl-thiobenzamide,benzenecarbothioamide, 4-trifluoromethyl,4-trifluoromethyl thiobenzamine
2734823
4-(trifluoromethyl)benzenecarbothioamide
C1=CC(=CC=C1C(=S)N)C(F)(F)F
72505-21-6
C8H6F3NS
205.2
IPRFNMJROWWFBH-UHFFFAOYSA-N
4-trifluoromethyl thiobenzamide,4-trifluoromethyl benzenecarbothioamide,4-trifluoromethylthiobenzamide,4-trifluoromethyl benzene-1-carbothioamide,4-trifluoromethyl benzothioamide,4-trifluoromethyl-thiobenzamide,benzenecarbothioamide, 4-trifluoromethyl,4-trifluoromethyl thiobenzamine
2734823
4-(trifluoromethyl)benzenecarbothioamide
C1=CC(=CC=C1C(=S)N)C(F)(F)F
Melting Point 136°C to 137°C
Packaging Glass bottle
Quantity 1g
Assay Percent Range 97.5% min. (HPLC)
Linear Formula CF3C6H4CSNH2
Formula Weight 205.2
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)thiobenzamide