Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Trifluoromethylphenylboronic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
500mg
5g
Packaging:
Glass bottle
128796-39-4
C7H6BF3O2
189.93
MFCD00151855
ALMFIOZYDASRRC-UHFFFAOYSA-N
4-trifluoromethyl phenylboronic acid,4-trifluoromethylphenylboronic acid,4-trifluoromethyl benzeneboronic acid,4-trifluoromethyl phenyl boronic acid,4-trifluoromethylphenyl boronic acid,4-trifluoromethyl phenyl boranediol,4-boronobenzotrifluoride,p-trifluoromethyl phenylboronic acid,4-trifluoromethylphenyl-boronic acid
2734389
[4-(trifluoromethyl)phenyl]boronic acid
B(C1=CC=C(C=C1)C(F)(F)F)(O)O
128796-39-4
C7H6BF3O2
189.93
MFCD00151855
ALMFIOZYDASRRC-UHFFFAOYSA-N
4-trifluoromethyl phenylboronic acid,4-trifluoromethylphenylboronic acid,4-trifluoromethyl benzeneboronic acid,4-trifluoromethyl phenyl boronic acid,4-trifluoromethylphenyl boronic acid,4-trifluoromethyl phenyl boranediol,4-boronobenzotrifluoride,p-trifluoromethyl phenylboronic acid,4-trifluoromethylphenyl-boronic acid
2734389
[4-(trifluoromethyl)phenyl]boronic acid
B(C1=CC=C(C=C1)C(F)(F)F)(O)O
Infrared Spectrum Authentic
Color White
Quantity 500mg
Melting Point 245.0°C to 250.0°C
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 189.93
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-Trifluoromethylphenylboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.