Alfa Aesar™ 4-(Trifluoromethylsulfonyl)benzonitrile, 97+%

Manufacturer: Alfa Aesar™ B2290706

Quantity 5g

 View more versions of this product

Catalog No. AAB2290706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 312-21-0
    Molecular Formula C8H4F3NO2S
    Formula Weight 235.18
    MDL Number MFCD01631631
    UN Number UN3439
    Synonym 4-(Trifluoromethanesulfonyl)benzonitrile
    Chemical Name or Material 4-(Trifluoromethylsulfonyl)benzonitrile

    Specifications

    Formula Weight 235.18
    Molecular Formula C8H4F3NO2S
    MDL Number MFCD01631631
    CAS 312-21-0
    UN Number UN3439
    Synonym 4-(Trifluoromethanesulfonyl)benzonitrile
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Melting Point 87° to 92°C
    Quantity 5g
    Health Hazard 1 Danger
    Chemical Name or Material 4-(Trifluoromethylsulfonyl)benzonitrile
    Assay ≥97%

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit