Alfa Aesar™ 4-(Trifluoromethylthio)benzylamine, 95%

Manufacturer: Alfa Aesar™ H3151503

Quantity 1g

 View more versions of this product

Catalog No. AAH3151503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 128273-56-3
    Molecular Formula C8H8F3NS
    Formula Weight 207.21
    MDL Number MFCD00190129
    UN Number UN2735
    Chemical Name or Material 4-(Trifluoromethylthio)benzylamine

    Specifications

    Formula Weight 207.21
    Molecular Formula C8H8F3NS
    MDL Number MFCD00190129
    CAS 128273-56-3
    UN Number UN2735
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Flash Point 80°C (176°F)
    Boiling Point 224° to 225°C
    Quantity 1g
    Chemical Name or Material 4-(Trifluoromethylthio)benzylamine
    Assay 95%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit