Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Trifluoromethylthio)phenol, 98%

Click to view available options
Quantity:
1g
5g
461-84-7
C7H5F3OS
194.171
MFCD00036035
YYCPTWHVKSATQK-UHFFFAOYSA-N
4-trifluoromethylthio phenol,4-trifluoromethyl thio phenol,4-trifluoromethyl sulfanyl phenol,4-trifluoromethylsulfanyl-phenol,4-trifluoromethylsulfanyl phenol,4-trifluoromethylthio hydroxybenzene,p-trifluoromethylthio phenol,4-trifluoromethyl sulphanyl phenol,phenol, 4-trifluoromethyl thio
3815158
4-(trifluoromethylsulfanyl)phenol
C1=CC(=CC=C1O)SC(F)(F)F
461-84-7
C7H5F3OS
194.171
MFCD00036035
YYCPTWHVKSATQK-UHFFFAOYSA-N
4-trifluoromethylthio phenol,4-trifluoromethyl thio phenol,4-trifluoromethyl sulfanyl phenol,4-trifluoromethylsulfanyl-phenol,4-trifluoromethylsulfanyl phenol,4-trifluoromethylthio hydroxybenzene,p-trifluoromethylthio phenol,4-trifluoromethyl sulphanyl phenol,phenol, 4-trifluoromethyl thio
3815158
4-(trifluoromethylsulfanyl)phenol
C1=CC(=CC=C1O)SC(F)(F)F
Quantity 1g
Melting Point 57°C to 60°C
Boiling Point 77°C to 78°C (7mmHg)
Formula Weight 194.18
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethylthio)phenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning